Methyl-3-nitrosalicylate ≥98%

Supplier: Thermo Fisher Scientific
Warning

B25029.06 B25029.14
B25029.06EA 119 EUR
B25029.06 B25029.14
Methyl-3-nitrosalicylate ≥98%
Methyl 3-nitrosalicylate

Purity: 98% CAS No.: 22621-41-6 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 129-132° MDL No.: MFCD00272263

Formula: C₈H₇NO₅
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00272263
CAS Number: 22621-41-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR