4-Azabenzimidazole 99%

Supplier: Thermo Fisher Scientific

B25051.03 B25051.06 B25051.14
B25051.03EA 66 EUR
B25051.03 B25051.06 B25051.14
4-Azabenzimidazole 99%
4-Azabenzimidazole

Purity: 99% CAS No.: 273-21-2 Molecular Formula: C6H5N3 Formula Weight: 119.13 Melting Point: 145-150° Boiling Point: 140-150°/0.1mm Beilstein Reference: 2600 MDL No.: MFCD00005579

Formula: C₆H₅N₃
Boiling Pt: 140…150 °C (0,1 mmHg)
Melting Pt: 145…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00005579
CAS Number: 273-21-2
EINECS: 205-987-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR