3-Amino-4-chloro-N,N-dimethylbenzamide 97%

Supplier: Thermo Fisher Scientific
Warning

B25294.06 B25294.14 B25294.22
B25294.06EA 26 EUR
B25294.06 B25294.14 B25294.22
3-Amino-4-chloro-N,N-dimethylbenzamide 97%
3-Amino-4-chloro-N,N-dimethylbenzamide

Purity: 97% CAS No.: 19694-10-1 Molecular Formula: C7H7ClN2O Formula Weight: 170.60 Melting Point: 164-166° MDL No.: MFCD00035785

Formula: C₇H₇ClN₂O
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00035785
CAS Number: 19694-10-1
EINECS: 243-231-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR