2-Bromo-5-fluorobenzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

B25394.03 B25394.06 B25394.14
B25394.03EA 0 EUR
B25394.03 B25394.06 B25394.14
2-Bromo-5-fluorobenzoic acid ≥98%
2-Bromo-5-fluorobenzoic acid

Purity: 98+% CAS No.: 394-28-5 Molecular Formula: C7H4BrFO2 Formula Weight: 219.01 Melting Point: 154-158° Beilstein Reference: 2575978

Formula: C₇H₄BrFO₂
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00142874
CAS Number: 394-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR