3,5-Dibromobenzaldehyde ≥98%

Supplier: Thermo Fisher Scientific
Warning

B25495.03 B25495.06 B25495.14
B25495.03EA 0 EUR
B25495.03 B25495.06 B25495.14
3,5-Dibromobenzaldehyde ≥98%
3,5-Dibromobenzaldehyde

Purity: 98% CAS No.: 56990-02-4 Molecular Formula: C7H4Br2O Formula Weight: 263.93 Melting Point: 89-93° Beilstein Reference: 2573432

Formula: C₇H₄Br₂O
MW: 263.92 g/mol
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00156887
CAS Number: 56990-02-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR