1-Aminopyridinium iodide 97%

Supplier: Thermo Fisher Scientific
Warning

B25703.06 B25703.14
B25703.06EA 57 EUR
B25703.06 B25703.14
1-Aminopyridinium iodide 97%
1-Aminopyridinium iodide

Purity: 97% CAS No.: 6295-87-0 Molecular Formula: C5H7IN2 Formula Weight: 222.03 Melting Point: ca 160° dec. Beilstein Reference: 3713241

Formula: C₅H₇IN₂
Melting Pt: ∼ 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051893
CAS Number: 6295-87-0
EINECS: 228-566-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR