5-Bromo-1-indanone ≥97%

Supplier: Thermo Fisher Scientific
Warning

B25707.03 B25707.06
B25707.03EA 42.4 EUR
B25707.03 B25707.06
5-Bromo-1-indanone ≥97%
5-Bromo-1-indanone

Purity: 98% CAS No.: 34598-49-7 Molecular Formula: C9H7BrO Formula Weight: 211.06 Melting Point: 125-129° Beilstein Reference: 2357199

Formula: C₉H₇BrO
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00082718
CAS Number: 34598-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR