Covers for Lab-Line® AquaBath® water baths, Lab-Line®
Supplier: Thermo Fisher Scientific
19000-11Q
BARN19000-11QEA 332 EUR
BARN19000-11Q
Covers for Lab-Line® AquaBath® water baths, Lab-Line®
Lids Bath Lids
Stainless steel covers ensure temperatures are maintained in Aquabath water baths.