IntelliStack™ Incubator Shakers, Single Unit
Supplier: Thermo Fisher Scientific
8882620D 8882620U 8885020D 8885020U
BARN8882620DEA 24600 EUR
BARN8882620D BARN8882620U BARN8885020D BARN8885020U
IntelliStack™ Incubator Shakers, Single Unit
Shakers Shaking Incubators
The Thermo Scientific™ IntelliStack™ Incubator shaker is a versatile laboratory instrument used for incubating and agitating samples simultaneously. It combines the functions of an incubator and shaker, providing a controlled environment for cell culture, microbial growth, and various other applications. With adjustable temperature, speed, and humidity settings, it offers precise control over experimental conditions. A large, easy-to-use 10" colour touchscreen display with real-time monitoring of key parameters and alarms, alerts, and data logging with export via USB or Ethernet allows easy
More Product Information