Screw filters for respirators
Supplier: BARTELS RIEGER
920600 920601 920603 920605 920850 920851 924400 924406 924700 924702 920650 924350
111-3072EA 10 EUR
111-3072 111-3073 111-3075 111-3076 111-3077 111-3078 111-3079 111-3081 111-3085 BART924702 BART920650 BART924350
Screw filters for respirators
Respirators Reusable Respirators Respirator Filters
Standardised respiratory filter.
  • Suitable for masks with standard EN 148-1 round-thread connections
EN 141, 143 and 371.