MBS (N-Succinimidyl 3-maleimidobenzoate)

Supplier: G Biosciences
Warning

BC11
BC11EA 443 EUR
BC11
MBS (N-Succinimidyl 3-maleimidobenzoate)
MBS (N-Succinimidyl 3-maleimidobenzoate)

Ideal for hapten-carrier protein, toxin-antibody, enzyme-antibody crosslinking

Formula: C₁₅H₁₀N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00005514
CAS Number: 58626-38-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a8b2e865b5ec39342a9929d1bd381a2dffed9ea&vid=7a8b2e865b5ec39342a9929d1bd381a2dffed9ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Spacer Arm (Å) 9.9
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable Yes
Water Soluble No
Cleavable/ Reversible No

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR