Beaker clip thermometer holder, multi-probe, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
B614007100
BELAB614007100EA 38.8 EUR
BELAB614007100
Beaker clip thermometer holder, multi-probe, SP Bel-Art
Thermometer Holders
The beaker clip liquid-in-glass thermometer holder can hold up to 4 thermometer probes with 3,5 to 4,0 mm ext.Ø at one time.
  • Clips easily onto the side of any beaker
  • Made from SUS304 stainless steel
Caution: WARNING–Cancer and Reproductive Harm. www.P65Warnings.ca.gov