Staining boxes, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F135511000
BELAF135511000EA 93.6 EUR
BELAF135511000
Staining boxes, SP Bel-Art
Staining Boxes
Boxes feature snap-fit lids, a drain (including an LD-PE plug) below the bottom of the box to virtually eliminate disturbance of the gel when liquid is being drained.
  • Liquid capacity up to 750 ml
  • Durable box and cover that are acid- and alkali-resistant
  • Also resistant to many solvents
Conveniently stain, fix, destain and manipulate electrophoresis gels.