Freezer storage box, 100 places, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F188510016
BELAF188510016EA 132 EUR
BELAF188510016
Freezer storage box, 100 places, SP Bel-Art
Boxes Cryoboxes
Freezer storage box provide compact, safe storage for tubes and vials.
  • Polypropylene construction
  • Available in a variety of colour choices
  • Hinged lids with snap tabs
  • Alphanumeric labeling
Great for organisation on the bench, in the freezer, or during transport, they can be used at temperatures –80 to +121 °C (112 to +249 °F)
Hinged lids with snap tab open easily and close securely, while clear translucent colour always lets you see inside. Opaque black boxes are ideal for light sensitive samples. Each box position is labeled with an alphanumericMore Product Information