Digital polarimeter, ADP450
Supplier: BELLINGHAM STANLEY
37-50
BELL37-50EA 16670 EUR
BELL37-50
Digital polarimeter, ADP450
Polarimeters
Digital polarimeter designed for measurements of specific rotation, percentage of concentration, purity, percentage of inverted sugar and percentage of inversion (A-B) in pharmaceutical, chemical, food and research applications.
  • Peltier temperature control
  • Standard and low volume sample tubes
  • Three decimal place resolution
  • High definition colour display
METHODS system makes convenient access to measurement types including continual measurement of specific rotation, optical rotation, concentration and inversion. MEAN-PHR Method allows a number ofMore Product Information