Neonate heads
Supplier: 3B Scientific
1005102 1005531
BINH1005102EA 1020 EUR
BINH1005102 BINH1005531
Neonate heads
Models Human Biology Models
This neonatal intubation trainer provides for practice of intubation skills on a newborn baby.
  • Realistic anatomy
  • Oral and nasal intubation, bag valve mask ventilation
  • Correct tube placement can be checked by practical inflation tests
Robust and realistic, this model allows students to undertake training that is directly transferable to the clinical setting.