Anti-SUGT1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB100250-100 ORB100250-20 ORB100250-200 ORB100250-5
BIRBORB100250-100EA 0 EUR
BIRBORB100250-100 BIRBORB100250-20 BIRBORB100250-200 BIRBORB100250-5
Anti-SUGT1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: SUGT1 (SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR