Anti-CDKN1A Rabbit polyclonal antibody

Supplier: Biorbyt
ORB107328-100
BIRBORB107328-100EA 0 EUR
BIRBORB107328-100
Anti-CDKN1A Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: CDKN1A (cyclin-dependent kinase inhibitor 1A (p21, Cip1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR