Anti-PH domain leucine-rich repeat-containing protein phosphatase 2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB109564-100
BIRBORB109564-100EA 0 EUR
BIRBORB109564-100
Anti-PH domain leucine-rich repeat-containing protein phosphatase 2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PH domain leucine-rich repeat-containing protein phosphatase 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR