Anti-PDCD1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Biorbyt
ORB114510-100
BIRBORB114510-100EA 0 EUR
BIRBORB114510-100
Anti-PDCD1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Type: Primary
Antigen: PDCD1 (programmed cell death 1)
Clonality: polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR