Anti-GPR55 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB124650-100
BIRBORB124650-100EA 0 EUR
BIRBORB124650-100
Anti-GPR55 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: GPR55 (G protein-coupled receptor 55)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR