Anti-SERPINA9 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)

Supplier: Biorbyt
ORB129158-100
BIRBORB129158-100EA 0 EUR
BIRBORB129158-100
Anti-SERPINA9 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)
Antibodies
Type: Primary
Antigen: SERPINA9 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR