Anti-NFATC4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB1302-100 ORB1302-200
BIRBORB1302-100EA 0 EUR
BIRBORB1302-100 BIRBORB1302-200
Anti-NFATC4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: NFATC4 (nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR