Anti-His tag Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)

Supplier: Biorbyt
ORB130693-100
BIRBORB130693-100EA 0 EUR
BIRBORB130693-100
Anti-His tag Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)
Antibodies
Type: Primary
Antigen: His tag (His tag)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol His
Environmentally Preferable
Antigen name His-tag
Antibody type Primary
Clonality Polyclonal
Conjugation PE (Phycoerythrin)/Cy7®
Reactivity
Host Rabbit
Isotype IgG
Immunogen C and N terminus
Sequence HHHHHH
Form liquid
Purification purified by affinity chromatography
Storage buffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR