Anti-SELL Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)

Supplier: Biorbyt
ORB130796-100
BIRBORB130796-100EA 0 EUR
BIRBORB130796-100
Anti-SELL Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)
Antibodies
Type: Primary
Antigen: SELL (selectin L)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR