Anti-SLAMF7 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)

Supplier: Biorbyt
ORB131118-100
BIRBORB131118-100EA 0 EUR
BIRBORB131118-100
Anti-SLAMF7 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)
Antibodies
Type: Primary
Antigen: SLAMF7 (SLAM family member 7)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR