Anti-SDHAF1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB137326-50
BIRBORB137326-50EA 0 EUR
BIRBORB137326-50
Anti-SDHAF1 Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 5 ?g/ml; Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: SDHAF1 (succinate dehydrogenase complex assembly factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR