Anti-GDF2 Goat Polyclonal Antibody

Supplier: Biorbyt
ORB19846-100
BIRBORB19846-100EA 0 EUR
BIRBORB19846-100
Anti-GDF2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GDF2 (growth differentiation factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR