Anti-APOL6 Goat Polyclonal Antibody

Supplier: Biorbyt
ORB20104-100
BIRBORB20104-100EA 0 EUR
BIRBORB20104-100
Anti-APOL6 Goat Polyclonal Antibody
Antibodies
ELISA: 1:8000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: APOL6 (apolipoprotein L, 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol APOL6
Environmentally Preferable
Antigen name Apolipoprotein L6
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype
Western blot Yes
ELISA Yes
Immunogen internal region
Molecular weight 38 kDa
Purification purified by ammonium sulphate precipitation followed by antigen affinity chromatography
Storage buffer Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR