Anti-JUNB Goat Polyclonal Antibody

Supplier: Biorbyt
ORB20240-100
BIRBORB20240-100EA 0 EUR
BIRBORB20240-100
Anti-JUNB Goat Polyclonal Antibody
Antibodies
ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: JUNB (jun B proto-oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR