Anti-NKX2-5 Goat Polyclonal Antibody

Supplier: Biorbyt
ORB20262-100
BIRBORB20262-100EA 0 EUR
BIRBORB20262-100
Anti-NKX2-5 Goat Polyclonal Antibody
Antibodies
ELISA: 1:32000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: NKX2-5 (NK2 homeobox 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR