Anti-Horse Albumin Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB22334-1
BIRBORB22334-1EA 0 EUR
BIRBORB22334-1
Anti-Horse Albumin Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Horse Albumin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR