Anti-Influenza A Goat Polyclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
ORB23143-1
BIRBORB23143-1EA 0 EUR
BIRBORB23143-1
Anti-Influenza A Goat Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Immunochemistry: 1:10-1:50

Type: Primary
Antigen: Influenza A
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR