Anti-TANC1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2991-100 ORB2991-200
BIRBORB2991-100EA 0 EUR
BIRBORB2991-100 BIRBORB2991-200
Anti-TANC1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: TANC1 (tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol TANC1
Antigen name tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen Synthetic peptide
Form liquid
Purification purified by Protein A chromatography
Concentration 0,5 mg/ml
Storage buffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR