Anti-CD72 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB43598-100
BIRBORB43598-100EA 0 EUR
BIRBORB43598-100
Anti-CD72 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: CD72 (CD72 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD72
Antigen name CD72 molecule
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Reactivity Human
Host Mouse
Isotype IgG2b
ImmunoPrecipitation Yes
Flow cytometry Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR