Anti-RBBP4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48521-100
BIRBORB48521-100EA 0 EUR
BIRBORB48521-100
Anti-RBBP4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RBBP4 (retinoblastoma binding protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol RBBP4
Antigen name Retinoblastoma binding protein 4
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Purification purified by affinity chromatography
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR