Anti-DPP9 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
ORB5025-100
BIRBORB5025-100EA 0 EUR
BIRBORB5025-100
Anti-DPP9 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: DPP9 (dipeptidyl-peptidase 9)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR