Anti-CERS2 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB6298-100 ORB6298-200
BIRBORB6298-100EA 0 EUR
BIRBORB6298-100 BIRBORB6298-200
Anti-CERS2 Rabbit polyclonal antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CERS2 (ceramide synthase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR