Anti-CCR5 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB67200-100
BIRBORB67200-100EA 0 EUR
BIRBORB67200-100
Anti-CCR5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000-1:3000

Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR