Anti-CDK1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB69546-100
BIRBORB69546-100EA 0 EUR
BIRBORB69546-100
Anti-CDK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR