Anti-RACGAP1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB74028-100
BIRBORB74028-100EA 0 EUR
BIRBORB74028-100
Anti-RACGAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RACGAP1 (Rac GTPase activating protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR