Anti-CASP4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB74031-100
BIRBORB74031-100EA 0 EUR
BIRBORB74031-100
Anti-CASP4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CASP4 (caspase 4, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR