Anti-CKAP4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB74209-100
BIRBORB74209-100EA 0 EUR
BIRBORB74209-100
Anti-CKAP4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CKAP4 (cytoskeleton-associated protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR