Anti-SPARC Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB76099-100
BIRBORB76099-100EA 0 EUR
BIRBORB76099-100
Anti-SPARC Rabbit Polyclonal Antibody
Antibodies
0.1-0.5 ?g/ml

Type: Primary
Antigen: SPARC (secreted protein, acidic, cysteine-rich (osteonectin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR