Anti-ARG2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB76318-100
BIRBORB76318-100EA 0 EUR
BIRBORB76318-100
Anti-ARG2 Rabbit Polyclonal Antibody
Antibodies
0.1-0.5 ?g/ml

Type: Primary
Antigen: ARG2 (arginase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR