Anti-ABCG5 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB77520-100
BIRBORB77520-100EA 0 EUR
BIRBORB77520-100
Anti-ABCG5 Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:5000-1:10000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: ABCG5 (ATP-binding cassette, sub-family G (WHITE), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR