Anti-IYD Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB78087-100
BIRBORB78087-100EA 0 EUR
BIRBORB78087-100
Anti-IYD Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:20000-1:80000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: IYD (iodotyrosine deiodinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR