Anti-E7 protein of human Papillomavirus type 11 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB79455-1
BIRBORB79455-1EA 0 EUR
BIRBORB79455-1
Anti-E7 protein of human Papillomavirus type 11 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: E7 protein of human Papillomavirus type 11
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR