Anti-RTN4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB88392-50
BIRBORB88392-50EA 0 EUR
BIRBORB88392-50
Anti-RTN4 Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 3 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: RTN4 (reticulon 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR