Anti-FAS Goat Polyclonal Antibody

Supplier: Biorbyt
ORB89673-50
BIRBORB89673-50EA 0 EUR
BIRBORB89673-50
Anti-FAS Goat Polyclonal Antibody
Antibodies
ELISA: 1:32000; Immunochemistry: 3.75 ?g/ml; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR